Assay Details
Target Gene Details
Entrez Gene ID: | 9891 |
Gene Name: | NUAK family kinase 1 |
Gene Aliases: |
ARK5 |
Location: |
Chr.12:106063347-106140033 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NUAK1 | NM_014840.2 | 7 | 3439 | NP_055655.1 |
| AB011109.1 | 7 | 3439 | ||
| AK027302.1 | BAB55026.1 | |||
| AK289992.1 | 7 | 2119 | ||
| BC152462.1 | 7 | 3439 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832509 | Chr.12:105970110 - 106122443 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map