Assay Details
Target Gene Details
Entrez Gene ID: | 94032 |
Gene Name: | calcium/calmodulin dependent protein kinase II inhibitor 2 |
Gene Aliases: |
CAM-KIIN, CAMKIIN |
Location: |
Chr.3:184259215-184261463 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAMK2N2 | NM_033259.2 | 2 | 1088 | NP_150284.1 |
Target Gene Details
Entrez Gene ID: | 9718 |
Gene Name: | endothelin converting enzyme 2 |
Gene Aliases: |
- |
Location: |
Chr.3:184249657-184293031 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ECE2 | NM_014693.3 | NP_055508.3 | ||
| AF428263.1 | AAL30386.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528639 | Chr.3:184228486 - 184262628 on Build GRCh38 | Loss |
|
| nsv592666 | Chr.3:184220630 - 184288607 on Build GRCh38 | Loss |
|
| nsv829812 | Chr.3:184136655 - 184302302 on Build GRCh38 | Loss |
|
| nsv4146 | Chr.3:184252606 - 184285216 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map