Assay Details
Target Gene Details
Entrez Gene ID: | 617 |
Gene Name: | BCS1 homolog, ubiquinol-cytochrome c reductase complex chaperone |
Gene Aliases: |
BCS, BCS1, BJS, FLNMS, GRACILE, Hs.6719, MC3DN1, PTD, h-BCS, h-BCS1 |
Location: |
Chr.2:218659656-218663443 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCS1L | NM_001079866.1 | 4 | 715 | NP_001073335.1 |
| NM_001257342.1 | 5 | 845 | NP_001244271.1 | |
| NM_001257343.1 | 5 | 897 | NP_001244272.1 | |
| NM_001257344.1 | 4 | 772 | NP_001244273.1 | |
| NM_001318836.1 | 3 | 346 | NP_001305765.1 | |
| NM_001320717.1 | 5 | 887 | NP_001307646.1 | |
| NM_004328.4 | 5 | 924 | NP_004319.1 | |
| XM_005246748.2 | 4 | 1199 | XP_005246805.1 | |
| XM_005246749.3 | 2 | 198 | XP_005246806.1 | |
| XM_006712678.1 | 4 | 1330 | XP_006712741.1 | |
| XM_017004631.1 | 5 | 1086 | XP_016860120.1 | |
| XM_017004632.1 | 5 | 811 | XP_016860121.1 | |
| XM_017004633.1 | 4 | 1589 | XP_016860122.1 | |
| XM_017004634.1 | 3 | 617 | XP_016860123.1 | |
| AF026849.1 | 4 | 691 | AAD08638.1 | |
| AF038195.1 | 4 | 653 | AAB97365.1 | |
| AK096210.1 | 4 | 1278 | ||
| AK223518.1 | 4 | 801 | BAD97238.1 | |
| AK289363.1 | 5 | 787 | ||
| BC000416.2 | 4 | 845 | AAH00416.1 | |
| BC007500.2 | 4 | 845 | AAH07500.1 | |
| BP215082.1 | 3 | 345 | ||
| BX571752.1 | 5 | 844 | CAE11877.1 | |
| KP992846.1 | 5 | 844 | ||
| KU177941.1 | 3 | 578 | ||
| KU177942.1 | 2 | 438 |
More Information
Set Membership: |
|

Genomic Map