Assay Details
Target Gene Details
Entrez Gene ID: | 7915 |
Gene Name: | aldehyde dehydrogenase 5 family member A1 |
Gene Aliases: |
SSADH, SSDH |
Location: |
Chr.6:24494969-24537207 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 1 - Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ALDH5A1 | NM_001080.3 | NP_001071.1 | ||
| NM_170740.1 | NP_733936.1 | |||
| AJ427353.1 | CAD20882.1 | |||
| AJ427354.2 | CAD20883.2 | |||
| AJ427355.2 | CAD20884.1 | |||
| AK290800.1 | ||||
| AK294699.1 | ||||
| AK314357.1 | ||||
| AK315380.1 | ||||
| BC034321.1 | AAH34321.1 | |||
| KJ534766.1 | ||||
| KJ534767.1 | ||||
| KJ534768.1 | ||||
| KJ534769.1 | ||||
| Y11192.1 | CAA72076.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv34020 | Chr.6:24481071 - 24605808 on Build GRCh38 | Loss |
|
| nsv519552 | Chr.6:24451028 - 24524476 on Build GRCh38 | Gain |
|
| esv2763542 | Chr.6:24444903 - 24520409 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map