Assay Details
Target Gene Details
Entrez Gene ID: | 1854 |
Gene Name: | deoxyuridine triphosphatase |
Gene Aliases: |
dUTPase |
Location: |
Chr.15:48331018-48343373 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUT | NM_001025248.1 | 7 | 1556 | NP_001020419.1 |
| NM_001025249.1 | 7 | 1240 | NP_001020420.1 | |
| NM_001948.3 | 6 | 1284 | NP_001939.1 | |
| AB049113.2 | 6 | 1219 | BAB13724.1 | |
| AK291515.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3636412 | Chr.15:48322023 - 48347079 on Build GRCh38 | Gain |
|
| esv2760030 | Chr.15:48216841 - 48371581 on Build GRCh38 | Loss |
|
| esv3569565 | Chr.15:48317122 - 48346517 on Build GRCh38 | Gain |
|
| nsv1053885 | Chr.15:48314149 - 48346517 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map