Assay Details
Target Gene Details
Entrez Gene ID: | 51527 |
Gene Name: | GSK3B interacting protein |
Gene Aliases: |
C14orf129, HSPC210 |
Location: |
Chr.14:96363452-96387290 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
GSKIP | NM_001271904.1 | 4 | 1014 | NP_001258833.1 |
NM_001271905.1 | 4 | 567 | NP_001258834.1 | |
NM_001271906.1 | 3 | 499 | NP_001258835.1 | |
NM_016472.4 | 4 | 600 | NP_057556.2 | |
XM_017021365.1 | 4 | 708 | XP_016876854.1 | |
AF151044.1 | 4 | 518 | AAF36130.1 | |
AK000796.1 | 4 | 526 | BAA91380.1 | |
AK094654.1 | 4 | 1016 | ||
AK314423.1 | 4 | 526 | ||
BC004818.2 | 3 | 486 | AAH04818.1 | |
DA488110.1 | ||||
HY108584.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
Target Variation |
Location | CNV Subtype |
Genes |
---|---|---|---|
esv3635423 | Chr.14:96385068 - 96403803 on Build GRCh38 | Gain |
![]() ![]() |
nsv565649 | Chr.14:96377253 - 96453283 on Build GRCh38 | Gain |
![]() ![]() |
