Assay Details
Target Gene Details
Entrez Gene ID: | 10826 |
Gene Name: | fatty acid hydroxylase domain containing 2 |
Gene Aliases: |
C5orf4 |
Location: |
Chr.5:154817018-154850685 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAXDC2 | NM_032385.4 | 9 | 2373 | NP_115761.2 |
| XM_005268358.2 | 6 | 2018 | XP_005268415.1 | |
| XM_006714753.2 | 10 | 2748 | XP_006714816.1 | |
| XM_017008965.1 | 10 | 2773 | XP_016864454.1 | |
| XM_017008966.1 | 7 | 2048 | XP_016864455.1 | |
| AF159165.1 | 1 | 2379 | AAF22611.1 | |
| AK023820.1 | 8 | 3164 | ||
| AK074245.1 | 3 | 1518 | ||
| AL359653.1 | 2 | 344 | ||
| AL359655.1 | 2 | 816 | ||
| BC004506.2 | 7 | 2070 | AAH04506.2 | |
| BC007216.1 | 9 | 2341 | AAH07216.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830521 | Chr.5:154768007 - 154864706 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77231999] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map