Assay Details
Target Gene Details
Entrez Gene ID: | 30817 |
Gene Name: | adhesion G protein-coupled receptor E2 |
Gene Aliases: |
CD312, EMR2, VBU |
Location: |
Chr.19:14732697-14778541 on Build GRCh38 |
Assay Gene Location: | Within Exon 24 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE2 | NM_001271052.1 | 19 | 2586 | NP_001257981.1 |
| NM_013447.3 | 21 | 3045 | NP_038475.2 | |
| AF114491.1 | 20 | 2658 | AAF21974.1 | |
| AK024426.1 | 11 | 1414 | BAB15716.1 | |
| AK024454.1 | 6 | 627 | BAB15744.1 | |
| AK291776.1 | 20 | 2791 | ||
| AK298700.1 | ||||
| BC034949.1 | 16 | 2792 | AAH34949.1 | |
| BC053933.1 | 16 | 2785 | AAH53933.1 | |
| BC127004.1 | 21 | 3041 | ||
| BC127005.1 | 21 | 3041 | ||
| CR936819.1 | 15 | 2022 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map