Assay Details
Target Gene Details
Entrez Gene ID: | 196951 |
Gene Name: | family with sequence similarity 227 member B |
Gene Aliases: |
C15orf33 |
Location: |
Chr.15:49326997-49620931 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
Target Gene Details
Entrez Gene ID: | 2252 |
Gene Name: | fibroblast growth factor 7 |
Gene Aliases: |
HBGF-7, KGF |
Location: |
Chr.15:49423178-49487326 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FGF7 | NM_002009.3 | NP_002000.1 | ||
| AK313160.1 | ||||
| BC010956.1 | 2 | 724 | AAH10956.1 | |
| CR542002.1 | CAG46799.1 | |||
| M60828.1 | AAA63210.1 | |||
| S81661.1 | AAB21431.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833002 | Chr.15:49329203 - 49489602 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map