Assay Details
Target Gene Details
Entrez Gene ID: | 8894 |
Gene Name: | eukaryotic translation initiation factor 2 subunit beta |
Gene Aliases: |
EIF2, EIF2B, EIF2beta, PPP1R67, eIF-2-beta |
Location: |
Chr.20:34088298-34112356 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF2S2 | NM_001316363.1 | 9 | 2190 | NP_001303292.1 |
| NM_001316364.1 | 8 | 2142 | NP_001303293.1 | |
| NM_003908.4 | 9 | 2199 | NP_003899.2 | |
| BX648379.1 | 2 | 1369 |
More Information
Set Membership: |
|

Genomic Map