Assay Details
Target Gene Details
Entrez Gene ID: | 480 |
Gene Name: | ATPase Na+/K+ transporting subunit alpha 4 |
Gene Aliases: |
ATP1A1, ATP1AL2 |
Location: |
Chr.1:160151562-160186977 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP1A4 | NM_144699.3 | 17 | 3020 | NP_653300.2 |
| XM_011509582.1 | 16 | 2487 | XP_011507884.1 | |
| AF506797.1 | 17 | 3012 | AAQ07964.1 | |
| AK098076.1 | 7 | 897 | BAC05228.1 | |
| AL833335.1 | 17 | 2993 | ||
| BC094801.1 | 17 | 2971 | AAH94801.1 | |
| BX647234.1 | 17 | 2994 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761950 | Chr.1:160158328 - 160193847 on Build GRCh38 | Gain |
|
| nsv522918 | Chr.1:160153991 - 160180340 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map