Assay Details
Target Gene Details
Entrez Gene ID: | 5481 |
Gene Name: | peptidylprolyl isomerase D |
Gene Aliases: |
CYP-40, CYPD |
Location: |
Chr.4:158709127-158723400 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PPID | NM_005038.2 | NP_005029.1 | ||
| AK313929.1 | ||||
| BC030707.2 | AAH30707.1 | |||
| BG699755.1 | ||||
| CR542081.1 | CAG46878.1 | |||
| L11667.1 | AAA35731.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830125 | Chr.4:158691163 - 158848360 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map