Assay Details
Target Gene Details
Entrez Gene ID: | 6775 |
Gene Name: | signal transducer and activator of transcription 4 |
Gene Aliases: |
SLEB11 |
Location: |
Chr.2:191029576-191172684 on Build GRCh38 |
Assay Gene Location: | Within Exon 27 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT4 | NM_001243835.1 | 21 | 2191 | NP_001230764.1 |
| NM_003151.3 | 21 | 2242 | NP_003142.1 | |
| XM_006712719.3 | 21 | 2237 | XP_006712782.1 | |
| XM_011511705.2 | 24 | 2388 | XP_011510007.1 | |
| AB209043.1 | 11 | 878 | BAD92280.1 | |
| AK299976.1 | 18 | 1723 | ||
| AK300868.1 | 21 | 1995 | ||
| AK301172.1 | 19 | 1666 | ||
| BC031212.1 | 21 | 2163 | AAH31212.1 | |
| L78440.1 | 21 | 2008 | AAB05605.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv999391 | Chr.2:190884309 - 191057205 on Build GRCh38 | Gain |
|
| nsv834493 | Chr.2:190946647 - 191109514 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map