Assay Details
Target Gene Details
Entrez Gene ID: | 168400 |
Gene Name: | DEAD-box helicase 53 |
Gene Aliases: |
CAGE, CT26 |
Location: |
Chr.X:22999961-23002089 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DDX53 | NM_182699.3 | 1 | 830 | NP_874358.2 |
| AY039237.1 | 1 | 835 | AAK72102.1 | |
| BC067878.1 | 1 | 821 | AAH67878.1 |
Target Gene Details
Entrez Gene ID: | 100873065 |
Gene Name: | PTCHD1 antisense RNA (head to head) |
Gene Aliases: |
DDX53-AS1, PTCHD1AS1, PTCHD1AS2 |
Location: |
Chr.X:22193005-23293146 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTCHD1-AS | NR_073010.2 | |||
| BX115199.1 | ||||
| DA355362.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3573648 | Chr.X:22953327 - 23080815 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map