Assay Details
Target Gene Details
Entrez Gene ID: | 6776 |
Gene Name: | signal transducer and activator of transcription 5A |
Gene Aliases: |
MGF, STAT5 |
Location: |
Chr.17:42287547-42311943 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STAT5A | NM_001288718.1 | NP_001275647.1 | ||
| NM_001288719.1 | NP_001275648.1 | |||
| NM_001288720.1 | NP_001275649.1 | |||
| NM_003152.3 | NP_003143.2 | |||
| XM_005257624.3 | XP_005257681.1 | |||
| AB208972.1 | BAD92209.1 | |||
| AK291650.1 | ||||
| AK301457.1 | ||||
| BC027036.1 | AAH27036.1 | |||
| DQ471288.1 | ||||
| L41142.1 | AAA73962.1 | |||
| U43185.1 | AAB06589.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833449 | Chr.17:42274854 - 42421081 on Build GRCh38 | Loss |
|
| nsv1056904 | Chr.17:42025823 - 42363204 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map