Assay Details
Target Gene Details
Entrez Gene ID: | 10026 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class K |
Gene Aliases: |
GPI8 |
Location: |
Chr.1:77088981-77219447 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGK | NM_005482.2 | 11 | 3308 | NP_005473.1 |
| AK055071.1 | 1 | 927 | ||
| BX648473.1 | 8 | 3010 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv830292 | Chr.1:76959886 - 77118622 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map