Assay Details
Target Gene Details
Entrez Gene ID: | 6837 |
Gene Name: | mediator complex subunit 22 |
Gene Aliases: |
MED24, SURF5, surf-5 |
Location: |
Chr.9:133340901-133348156 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED22 | NM_133640.4 | NP_598395.1 | ||
| NM_181491.2 | 4 | 781 | NP_852468.1 | |
| AJ224358.1 | 4 | 726 | CAA11915.1 | |
| AJ224359.1 | CAA11916.1 | |||
| AK124518.1 | ||||
| AK126069.1 | 4 | 762 | ||
| BC024225.2 | 4 | 724 | AAH24225.1 | |
| BC040111.1 | 4 | 731 | AAH40111.1 | |
| BI756582.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2739138 | Chr.9:132727483 - 135521553 on Build GRCh38 | Deletion |
|
| esv3621939 | Chr.9:133330867 - 133344894 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map