Assay Details
Target Gene Details
Entrez Gene ID: | 84733 |
Gene Name: | chromobox 2 |
Gene Aliases: |
CDCA6, M33, SRXY5 |
Location: |
Chr.17:79776254-79787650 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CBX2 | NM_005189.2 | 5 | 495 | NP_005180.1 |
| XM_011525382.1 | 5 | 535 | XP_011523684.1 | |
| XM_011525383.2 | 5 | 1497 | XP_011523685.1 | |
| CN367085.1 | ||||
| CN367087.1 | 5 | 419 | ||
| CV815254.1 | 5 | 396 | ||
| DB452048.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv538n67 | Chr.17:79765152 - 79848648 on Build GRCh38 | Gain |
|
| nsv469865 | Chr.17:79770348 - 79957778 on Build GRCh38 | Loss |
|
| nsv828116 | Chr.17:79748992 - 79855333 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map