Assay Details
Target Gene Details
Entrez Gene ID: | 10362 |
Gene Name: | high mobility group 20B |
Gene Aliases: |
BRAF25, BRAF35, HMGX2, HMGXB2, PP7706, SMARCE1r, SOXL, pp8857 |
Location: |
Chr.19:3572938-3579083 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HMG20B | NM_006339.2 | NP_006330.2 | ||
| XM_017026144.1 | XP_016881633.1 | |||
| AF072165.1 | AAF76253.1 | |||
| AF072836.1 | AAC26860.1 | |||
| AF146223.1 | AAF66707.1 | |||
| AF288679.1 | AAG01174.1 | |||
| AF318366.1 | AAL55873.1 | |||
| AF331191.1 | AAG60060.1 | |||
| AK090733.1 | BAC03510.1 | |||
| AK129638.1 | ||||
| AK289500.1 | ||||
| AK314201.1 | ||||
| AL355691.1 | ||||
| AL355702.1 | ||||
| AL355703.1 | ||||
| AL355709.1 | CAB90809.2 | |||
| BC002552.2 | AAH02552.1 | |||
| BC003505.1 | AAH03505.2 | |||
| BC004408.1 | AAH04408.2 | |||
| BC021585.1 | AAH21585.1 | |||
| CR456754.1 | CAG33035.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv961165 | Chr.19:3567351 - 3579402 on Build GRCh38 | Duplication |
|
| nsv953948 | Chr.19:3171703 - 3700002 on Build GRCh38 | Deletion |
|
| nsv833713 | Chr.19:3564423 - 3627361 on Build GRCh38 | Loss |
|
| nsv470114 | Chr.19:3372516 - 3855979 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map