Assay Details
Target Gene Details
Entrez Gene ID: | 599 |
Gene Name: | BCL2 like 2 |
Gene Aliases: |
BCL-W, BCL2-L-2, BCLW, PPP1R51 |
Location: |
Chr.14:23306762-23311759 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L2 | NM_001199839.1 | 4 | 3268 | NP_001186768.1 |
| NM_004050.4 | 4 | 3313 | NP_004041.1 | |
| AA524030.1 | 1 | 298 | ||
| AA741032.1 | 1 | 294 | ||
| BQ001111.1 | 1 | 309 | ||
| D87461.1 | 4 | 3260 |
Target Gene Details
Entrez Gene ID: | 100529063 |
Gene Name: | BCL2L2-PABPN1 readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:23306762-23326185 on Build GRCh38 |
Assay Gene Location: | Within Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BCL2L2-PABPN1 | NM_001199864.1 | NP_001186793.1 |
More Information
Set Membership: |
|

Genomic Map