Assay Details
Target Gene Details
| Entrez Gene ID: | 10717 | 
| Gene Name: | adaptor related protein complex 4 beta 1 subunit | 
| Gene Aliases: | BETA-4, CPSQ5, SPG47 | 
| Location: | Chr.1:113894194-113905202 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 11 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| AP4B1 | NM_001253852.2 | 10 | 2077 | NP_001240781.1 | 
| NM_001253853.2 | 11 | 2002 | NP_001240782.1 | |
| NM_001308312.1 | 7 | 1573 | NP_001295241.1 | |
| NM_006594.4 | 11 | 2130 | NP_006585.2 | |
| XM_011540523.2 | 10 | 1983 | XP_011538825.1 | |
| XM_011540524.2 | 9 | 1817 | XP_011538826.1 | |
| XM_011540525.2 | 9 | 1929 | XP_011538827.1 | |
| XM_011540528.2 | 10 | 4835 | XP_011538830.1 | |
| XM_017000088.1 | 10 | 3814 | XP_016855577.1 | |
| XM_017000089.1 | 8 | 1613 | XP_016855578.1 | |
| XM_017000090.1 | 8 | 1568 | XP_016855579.1 | |
| XM_017000091.1 | 7 | 1617 | XP_016855580.1 | |
| XM_017000092.1 | 11 | 2194 | XP_016855581.1 | |
| AB209817.1 | 10 | 2014 | BAD93054.1 | |
| AF092094.1 | 11 | 2074 | AAD20448.1 | |
| AK001045.1 | 4 | 1582 | ||
| AK057567.1 | 2 | 2313 | ||
| AK093740.1 | 7 | 1478 | ||
| AK300204.1 | 11 | 1841 | ||
| AK314641.1 | 11 | 2034 | ||
| BC014146.2 | 10 | 1935 | AAH14146.1 | 
Target Gene Details
| Entrez Gene ID: | 100287722 | 
| Gene Name: | AP4B1 antisense RNA 1 | 
| Gene Aliases: | - | 
| Location: | Chr.1:113812612-113901237 on Build GRCh38 | 
| Assay Gene Location: | Within Intron 6 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| AP4B1-AS1 | NR_037864.1 | |||
| NR_125965.1 | ||||
| AK123199.1 | ||||
| DB028504.1 | ||||
| DB074478.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv3587251 | Chr.1:113871703 - 113936287 on Build GRCh38 | Loss |  DCLRE1B  HIPK1  AP4B1  BCL2L15  PTPN22  AP4B1-AS1  HIPK1-AS1 | 
| esv3587240 | Chr.1:113533246 - 114158910 on Build GRCh38 | Loss |  DCLRE1B  PHTF1  OLFML3  RSBN1  HIPK1  SYT6  MAGI3  AP4B1  BCL2L15  PTPN22  AP4B1-AS1  HIPK1-AS1 | 
| esv23869 | Chr.1:113320330 - 114358495 on Build GRCh38 | Loss |  DCLRE1B  PHTF1  OLFML3  RSBN1  HIPK1  SYT6  MAGI3  AP4B1  BCL2L15  PTPN22  AP4B1-AS1  HIPK1-AS1 | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Intronic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top