Assay Details
Target Gene Details
Entrez Gene ID: | 285672 |
Gene Name: | SREK1 interacting protein 1 |
Gene Aliases: |
P18SRP, SFRS12IP1 |
Location: |
Chr.5:64718148-64768849 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SREK1IP1 | NM_173829.3 | 5 | 4136 | NP_776190.1 |
| XM_011543344.2 | 4 | 4268 | XP_011541646.1 | |
| AL832348.1 | 5 | 4093 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv5692n100 | Chr.5:64578799 - 64749903 on Build GRCh38 | Gain |
|
| nsv598390 | Chr.5:64554330 - 64982002 on Build GRCh38 | Gain |
|
| esv2752074 | Chr.5:64636623 - 64733547 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map