Assay Details
Target Gene Details
Entrez Gene ID: | 390260 |
Gene Name: | olfactory receptor family 6 subfamily X member 1 |
Gene Aliases: |
OR11-270 |
Location: |
Chr.11:123753580-123754518 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OR6X1 | NM_001005188.1 | 1 | 118 | NP_001005188.1 |
| BC136855.1 | 1 | 145 | ||
| BC136856.1 | 1 | 145 |
More Information
Set Membership: |
|

Genomic Map