Assay Details
Target Gene Details
Entrez Gene ID: | 9836 |
Gene Name: | leucine carboxyl methyltransferase 2 |
Gene Aliases: |
PPM2, TYW4 |
Location: |
Chr.15:43327776-43330622 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCMT2 | NM_014793.4 | 1 | 1612 | NP_055608.2 |
| AF265443.1 | 1 | 1105 | AAF75774.1 | |
| AK292587.1 | 1 | 1538 | ||
| AK300820.1 | 2 | 821 | ||
| BC015949.2 | 1 | 1595 | AAH15949.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1512 | Chr.15:43324636 - 43369448 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map