Assay Details
Target Gene Details
Entrez Gene ID: | 128338 |
Gene Name: | DNA damage regulated autophagy modulator 2 |
Gene Aliases: |
CORD21, PRO180, TMEM77, WWFQ154 |
Location: |
Chr.1:111117332-111140216 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRAM2 | NM_178454.4 | 9 | 1600 | NP_848549.3 |
| XM_005270469.2 | 10 | 1658 | XP_005270526.1 | |
| XM_005270470.1 | 10 | 1781 | XP_005270527.1 | |
| XM_006710361.1 | 10 | 1618 | XP_006710424.1 | |
| XM_006710362.1 | 10 | 1591 | XP_006710425.1 | |
| XM_011540707.1 | 10 | 1599 | XP_011539009.1 | |
| XM_011540708.1 | 10 | 1679 | XP_011539010.1 | |
| XM_017000320.1 | 9 | 1542 | XP_016855809.1 | |
| XM_017000321.1 | 9 | 1401 | XP_016855810.1 | |
| AK075350.1 | 5 | 1046 | BAC11562.1 | |
| AY336747.1 | 9 | 1478 | AAR02410.1 | |
| BQ006335.1 | 1 | 302 |
More Information
Additional Information:
For this assay, SNP(s) [rs112279674] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map