Assay Details
Target Gene Details
Entrez Gene ID: | 8815 |
Gene Name: | barrier to autointegration factor 1 |
Gene Aliases: |
BAF, BCRP1, D14S1460, NGPS |
Location: |
Chr.11:66002079-66004149 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BANF1 | NM_001143985.1 | 1 | 155 | NP_001137457.1 |
| NM_003860.3 | 1 | 155 | NP_003851.1 | |
| AF068235.1 | 1 | 155 | AAD15901.1 | |
| BU940708.1 |
Target Gene Details
Entrez Gene ID: | 84285 |
Gene Name: | eukaryotic translation initiation factor 1A domain containing |
Gene Aliases: |
haponin |
Location: |
Chr.11:65996545-66002217 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EIF1AD | XM_017018412.1 | XP_016873901.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832191 | Chr.11:65864858 - 66038649 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs78790365] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map