Assay Details
Target Gene Details
Entrez Gene ID: | 5269 |
Gene Name: | serpin family B member 6 |
Gene Aliases: |
CAP, DFNB91, MSTP057, PI-6, PI6, PTI, SPI3 |
Location: |
Chr.6:2948159-2972165 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SERPINB6 | NM_001195291.2 | 1 | 382 | NP_001182220.2 |
| NM_001271824.1 | 1 | 382 | NP_001258753.1 | |
| NM_001297699.1 | 1 | 382 | NP_001284628.1 | |
| NM_001297700.1 | 1 | 382 | NP_001284629.1 | |
| NM_004568.5 | 1 | 382 | NP_004559.4 | |
| XM_011514672.1 | XP_011512974.1 | |||
| AK094983.1 | 1 | 382 | ||
| AK314578.1 | 1 | 101 | ||
| BI827451.1 | 1 | 73 | ||
| BM789272.1 | ||||
| CD674931.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv600833 | Chr.6:2970334 - 3010915 on Build GRCh38 | Loss |
|
| nsv830575 | Chr.6:2847376 - 3035682 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map