Assay Details
Target Gene Details
| Entrez Gene ID: | 54487 | 
| Gene Name: | DGCR8 microprocessor complex subunit | 
| Gene Aliases: | C22orf12, DGCRK6, Gy1, pasha | 
| Location: | Chr.22:20080232-20111877 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 2 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| DGCR8 | NM_001190326.1 | 2 | 586 | NP_001177255.1 | 
| NM_022720.6 | 2 | 586 | NP_073557.3 | |
| AF165527.1 | 1 | 247 | AAF82263.1 | |
| AK313357.1 | 2 | 537 | ||
| BC009984.2 | 2 | 507 | AAH09984.1 | |
| BC078147.1 | 1 | 300 | AAH78147.1 | |
| BQ227899.1 | 2 | 543 | ||
| CR456356.1 | 1 | 244 | CAG30242.1 | |
| DA433651.1 | 
Target Gene Details
| Entrez Gene ID: | 100302197 | 
| Gene Name: | microRNA 1306 | 
| Gene Aliases: | MIRN1306, hsa-mir-1306, mir-1306 | 
| Location: | Chr.22:20086058-20086142 on Build GRCh38 | 
| Assay Gene Location: | Overlaps - Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MIR1306 | NR_031706.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| dgv726n67 | Chr.22:20070444 - 20324465 on Build GRCh38 | Gain |  DGCR6L  ZDHHC8  LOC440792  RANBP1  CCDC188  RTN4R  MIR6816  TRMT2A  DGCR8  LINC00896  MIR1286  MIR1306  MIR3618  LOC284865 | 
| esv3647284 | Chr.22:19985583 - 20137122 on Build GRCh38 | Gain |  DGCR8  ZDHHC8  ARVCF  MIR1306  RANBP1  MIR185  TANGO2  MIR3618  MIR6816  TRMT2A | 
| esv3893434 | Chr.22:19470889 - 20343372 on Build GRCh38 | Gain |  DGCR6L  ZDHHC8  ARVCF  CDC45  LOC440792  SEPT5-GP1BB  LINC00895  RTN4R  MIR4761  TRMT2A  TXNRD2  UFD1L  LINC00896  COMT  C22orf29  MIR3618  TBX1  RANBP1  SEPT5  CCDC188  MIR185  MIR6816  DGCR8  MIR1286  GP1BB  MIR1306  CLDN5  LOC101927859  TANGO2  LOC284865  GNB1L | 
| esv2758825 | Chr.22:20038970 - 20321924 on Build GRCh38 | Loss |  DGCR6L  ZDHHC8  LOC440792  RANBP1  CCDC188  RTN4R  MIR6816  TRMT2A  DGCR8  LINC00896  MIR1286  MIR1306  TANGO2  MIR3618  LOC284865 | 
| nsv828939 | Chr.22:19696032 - 20324465 on Build GRCh38 | Loss |  DGCR6L  ZDHHC8  ARVCF  LOC440792  SEPT5-GP1BB  RTN4R  MIR4761  TRMT2A  TXNRD2  LINC00896  COMT  C22orf29  MIR3618  TBX1  RANBP1  SEPT5  CCDC188  MIR185  MIR6816  DGCR8  MIR1286  GP1BB  MIR1306  TANGO2  LOC284865  GNB1L | 
| nsv828938 | Chr.22:19691069 - 20324465 on Build GRCh38 | Gain |  DGCR6L  ZDHHC8  ARVCF  LOC440792  SEPT5-GP1BB  RTN4R  MIR4761  TRMT2A  TXNRD2  LINC00896  COMT  C22orf29  MIR3618  TBX1  RANBP1  SEPT5  CCDC188  MIR185  MIR6816  DGCR8  MIR1286  GP1BB  MIR1306  TANGO2  LOC284865  GNB1L | 
| dgv4480n100 | Chr.22:20069620 - 20262667 on Build GRCh38 | Gain |  DGCR8  ZDHHC8  LINC00896  MIR1286  MIR1306  RANBP1  CCDC188  RTN4R  MIR3618  MIR6816  LOC284865  TRMT2A | 
| nsv471183 | Chr.22:20018517 - 20319470 on Build GRCh38 | Loss |  DGCR6L  ZDHHC8  LOC440792  RANBP1  CCDC188  MIR185  RTN4R  MIR6816  TRMT2A  DGCR8  LINC00896  MIR1286  MIR1306  TANGO2  MIR3618  LOC284865 | 
| nsv953027 | Chr.22:20085378 - 20303977 on Build GRCh38 | Deletion |  ZDHHC8  LOC440792  RANBP1  CCDC188  RTN4R  MIR6816  TRMT2A  DGCR8  LINC00896  MIR1286  MIR1306  MIR3618  LOC284865 | 
| nsv834133 | Chr.22:20029363 - 20176708 on Build GRCh38 | Loss |  DGCR8  ZDHHC8  MIR1306  RANBP1  CCDC188  MIR185  TANGO2  MIR3618  MIR6816  TRMT2A | 
| dgv7983n54 | Chr.22:19762002 - 20321870 on Build GRCh38 | Gain |  DGCR6L  ZDHHC8  ARVCF  TBX1  LOC440792  RANBP1  CCDC188  MIR185  RTN4R  MIR6816  MIR4761  TRMT2A  TXNRD2  DGCR8  LINC00896  MIR1286  COMT  MIR1306  C22orf29  TANGO2  MIR3618  LOC284865  GNB1L | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  Non-exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top