Assay Details
Target Gene Details
Entrez Gene ID: | 84260 |
Gene Name: | trichoplein keratin filament binding |
Gene Aliases: |
TpMs |
Location: |
Chr.12:109880667-109918069 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TCHP | NM_001143852.1 | NP_001137324.1 | ||
| NM_032300.4 | 1 | 90 | NP_115676.1 | |
| XM_011538836.2 | XP_011537138.1 | |||
| XM_011538837.2 | XP_011537139.1 | |||
| XM_017020024.1 | XP_016875513.1 | |||
| XM_017020025.1 | XP_016875514.1 | |||
| AK092736.1 | BAC03960.1 | |||
| AK301262.1 | ||||
| AY007230.1 | 1 | 90 | AAG12971.1 | |
| BC004285.1 | AAH04285.1 |
More Information
Set Membership: |
|

Genomic Map