Assay Details
Target Gene Details
Entrez Gene ID: | 84647 |
Gene Name: | phospholipase A2 group XIIB |
Gene Aliases: |
FKSG71, GXIIB, GXIIIsPLA2, PLA2G13, sPLA2-GXIIB |
Location: |
Chr.10:72934762-72955044 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G12B | NM_001318124.1 | 4 | 1103 | NP_001305053.1 |
| NM_001318125.1 | 3 | 1017 | NP_001305054.1 | |
| NM_032562.4 | 4 | 1106 | NP_115951.2 | |
| XM_005270244.4 | 4 | 1319 | XP_005270301.1 | |
| AF339053.1 | 4 | 1065 | AAL09472.1 | |
| AK311349.1 | 3 | 1004 | ||
| AY358032.1 | AAQ92918.1 | |||
| BF939574.1 | 1 | 54 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv528473 | Chr.10:72911853 - 73049838 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map