Assay Details
Target Gene Details
Entrez Gene ID: | 6648 |
Gene Name: | superoxide dismutase 2, mitochondrial |
Gene Aliases: |
IPO-B, IPOB, MNSOD, MVCD6, Mn-SOD |
Location: |
Chr.6:159679064-159762529 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SOD2 | NM_001322817.1 | NP_001309746.1 | ||
| AK097395.1 |
Target Gene Details
Entrez Gene ID: | 9589 |
Gene Name: | Wilms tumor 1 associated protein |
Gene Aliases: |
Mum2 |
Location: |
Chr.6:159726680-159756319 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WTAP | NM_001270531.1 | NP_001257460.1 | ||
| NM_001270532.1 | NP_001257461.1 | |||
| NM_001270533.1 | NP_001257462.1 | |||
| NM_004906.4 | NP_004897.2 | |||
| NM_152857.2 | NP_690596.1 | |||
| NM_152858.2 | NP_690597.1 | |||
| XM_017011514.1 | XP_016867003.1 | |||
| XM_017011516.1 | XP_016867005.1 | |||
| AF374416.1 | AAK54764.1 | |||
| AJ276706.1 | CAC10188.1 | |||
| AK127822.1 | ||||
| AK290854.1 | ||||
| BC000383.2 | AAH00383.1 | |||
| BC004432.1 | AAH04432.1 | |||
| BC069192.1 | AAH69192.1 | |||
| BE906617.1 | ||||
| BG702882.1 | ||||
| BQ948913.1 | ||||
| BX387163.2 | ||||
| CN284477.1 | ||||
| D14661.1 | BAA03495.1 | |||
| DA138013.1 | ||||
| DA561768.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv949879 | Chr.6:159733269 - 159742868 on Build GRCh38 | Duplication |
|
| esv2759483 | Chr.6:159582799 - 159794119 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map