Assay Details
Target Gene Details
Entrez Gene ID: | 1326 |
Gene Name: | mitogen-activated protein kinase kinase kinase 8 |
Gene Aliases: |
AURA2, COT, EST, ESTF, MEKK8, TPL2, Tpl-2, c-COT |
Location: |
Chr.10:30434021-30461833 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP3K8 | NM_001244134.1 | 2 | 544 | NP_001231063.1 |
| NM_001320961.1 | 2 | 300 | NP_001307890.1 | |
| NM_005204.3 | 3 | 775 | NP_005195.2 | |
| XM_017015707.1 | 2 | 992 | XP_016871196.1 | |
| XM_017015708.1 | 3 | 505 | XP_016871197.1 | |
| XM_017015709.1 | 2 | 3285 | XP_016871198.1 | |
| XM_017015710.1 | 2 | 395 | XP_016871199.1 | |
| XM_017015711.1 | 2 | 992 | XP_016871200.1 | |
| XM_017015712.1 | 3 | 505 | XP_016871201.1 | |
| XM_017015713.1 | 3 | 749 | XP_016871202.1 | |
| XM_017015714.1 | 2 | 544 | XP_016871203.1 | |
| AB209539.1 | 1 | 2902 | BAD92776.1 | |
| AK290320.1 | 2 | 440 | ||
| AK313295.1 | 2 | 441 | ||
| AL547407.3 | 3 | 866 | ||
| BC104833.1 | 2 | 401 | AAI04834.1 | |
| BC113566.1 | 2 | 401 | ||
| BI753824.1 | 2 | 270 | ||
| CR542284.1 | 1 | 163 | CAG47079.1 | |
| D14497.1 | 3 | 529 | BAA03387.1 | |
| DB271664.1 | ||||
| Z14138.1 | 1 | 323 | CAA78512.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1161936 | Chr.10:30113251 - 31078102 on Build GRCh38 | Duplication |
|
| nsv831821 | Chr.10:30346221 - 30536200 on Build GRCh38 | Loss |
|
More Information
Additional Information:
For this assay, SNP(s) [rs77186746] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map