Assay Details
Target Gene Details
Entrez Gene ID: | 55616 |
Gene Name: | ArfGAP with SH3 domain, ankyrin repeat and PH domain 3 |
Gene Aliases: |
ACAP4, CENTB6, DDEFL1, UPLC1 |
Location: |
Chr.1:23428563-23484631 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
ASAP3 | NM_001143778.1 | 13 | 1384 | NP_001137250.1 |
NM_017707.3 | 14 | 1411 | NP_060177.2 | |
XM_011541755.1 | 14 | 1351 | XP_011540057.1 | |
XM_017001685.1 | 14 | 1345 | XP_016857174.1 | |
XM_017001686.1 | 14 | 1351 | XP_016857175.1 | |
XM_017001687.1 | 14 | 1318 | XP_016857176.1 | |
XM_017001688.1 | 14 | 1307 | XP_016857177.1 | |
XM_017001689.1 | 14 | 1368 | XP_016857178.1 | |
AB056749.1 | 14 | 1332 | BAB85677.1 | |
AK000253.1 | 4 | 325 | BAA91033.1 | |
AK092291.1 | 14 | 1333 | ||
AK295100.1 | 13 | 1306 | ||
AK299359.1 | 14 | 1307 | ||
BC023519.2 | AAH23519.1 | |||
BC060786.1 | 14 | 1359 | AAH60786.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
