Assay Details
Target Gene Details
Entrez Gene ID: | 129684 |
Gene Name: | contactin associated protein like 5 |
Gene Aliases: |
caspr5 |
Location: |
Chr.2:124025287-124921201 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNTNAP5 | NM_130773.3 | 1 | 232 | NP_570129.1 |
| XM_006712258.1 | 1 | 232 | XP_006712321.1 | |
| XM_017003316.1 | 1 | 232 | XP_016858805.1 | |
| AK056528.1 | 1 | 232 | BAB71205.1 |
More Information
Set Membership: |
|

Genomic Map