Assay Details
Target Gene Details
Entrez Gene ID: | 5368 |
Gene Name: | prepronociceptin |
Gene Aliases: |
N/OFQ, NOP, OFQ, PPNOC, ppN/OFQ |
Location: |
Chr.8:28316979-28343355 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PNOC | NM_001284244.1 | 2 | 363 | NP_001271173.1 |
| NM_006228.4 | 3 | 556 | NP_006219.1 | |
| XM_005273532.2 | 3 | 643 | XP_005273589.1 | |
| XM_011544559.2 | 3 | 446 | XP_011542861.1 | |
| XM_017013578.1 | 2 | 1094 | XP_016869067.1 | |
| AK289633.1 | 3 | 419 | ||
| AK301456.1 | 2 | 363 | ||
| AY335948.1 | 3 | 418 | AAP94601.1 | |
| BC034758.1 | 3 | 401 | AAH34758.1 | |
| BF976583.1 | 2 | 343 | ||
| BT019334.1 | 2 | 348 | AAV38141.1 | |
| CR541940.1 | 2 | 348 | CAG46738.1 | |
| CR541965.1 | 2 | 348 | CAG46763.1 | |
| U48263.1 | 3 | 559 | AAC50651.1 | |
| X97370.1 | 2 | 371 | CAA66040.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv456n21 | Chr.8:28321496 - 28467894 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs76786693] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map