Assay Details
Target Gene Details
Entrez Gene ID: | 976 |
Gene Name: | adhesion G protein-coupled receptor E5 |
Gene Aliases: |
CD97, TM7LN1 |
Location: |
Chr.19:14381144-14408725 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 12 - Intron 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADGRE5 | NM_001025160.2 | NP_001020331.1 | ||
| NM_001784.4 | NP_001775.2 | |||
| NM_078481.3 | NP_510966.1 | |||
| XM_011528451.2 | XP_011526753.1 | |||
| XM_017027547.1 | XP_016883036.1 | |||
| AK097264.1 | ||||
| AK225655.1 | ||||
| AK225677.1 | ||||
| AK292159.1 | ||||
| AK300343.1 | ||||
| AK304553.1 | ||||
| AK308368.1 | ||||
| AK314697.1 | ||||
| AX775811.1 | CAE11605.1 | |||
| AX775813.1 | CAE11606.1 | |||
| BC026690.2 | AAH26690.1 | |||
| U76764.1 | AAB36682.1 | |||
| X84700.1 | CAA59173.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1160590 | Chr.19:14398750 - 14462842 on Build GRCh38 | Deletion |
|
| nsv833764 | Chr.19:14381787 - 14504061 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map