Assay Details
Target Gene Details
Entrez Gene ID: | 5655 |
Gene Name: | kallikrein related peptidase 10 |
Gene Aliases: |
NES1, PRSSL1 |
Location: |
Chr.19:51012744-51020175 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLK10 | NM_001077500.1 | NP_001070968.1 | ||
| NM_002776.4 | NP_002767.2 | |||
| NM_145888.2 | NP_665895.1 | |||
| XM_005259061.3 | XP_005259118.1 | |||
| XM_005259062.3 | XP_005259119.1 | |||
| XM_006723287.3 | XP_006723350.1 | |||
| XM_006723289.3 | XP_006723352.1 | |||
| XM_017026993.1 | XP_016882482.1 | |||
| AF024605.1 | AAB81602.1 | |||
| AK292365.1 | ||||
| AK309456.1 | ||||
| AY561635.1 | AAS66976.1 | |||
| BC002710.2 | AAH02710.1 | |||
| CR450327.1 | CAG29323.1 | |||
| KJ010190.1 | ||||
| KJ010191.1 | ||||
| KJ010192.1 | ||||
| KJ010193.1 | ||||
| KJ010194.1 | ||||
| KJ010195.1 | ||||
| KJ010196.1 | ||||
| KJ010197.1 | ||||
| KJ010198.1 | ||||
| KJ010199.1 | ||||
| KJ010200.1 | ||||
| KJ010201.1 | ||||
| KJ010202.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2677627 | Chr.19:51015389 - 51020436 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map