Assay Details
Target Gene Details
Entrez Gene ID: | 4176 |
Gene Name: | minichromosome maintenance complex component 7 |
Gene Aliases: |
CDC47, MCM2, P1.1-MCM3, P1CDC47, P85MCM, PNAS146, PPP1R104 |
Location: |
Chr.7:100092728-100101940 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCM7 | NM_001278595.1 | 14 | 2863 | NP_001265524.1 |
| NM_005916.4 | 15 | 2788 | NP_005907.3 | |
| NM_182776.2 | 14 | 2754 | NP_877577.1 | |
| XM_005250348.3 | 15 | 2442 | XP_005250405.1 | |
| XM_017012217.1 | 15 | 2319 | XP_016867706.1 | |
| AK055379.1 | 14 | 2731 | ||
| AK096959.1 | 16 | 2152 | ||
| AK226175.1 | 14 | 2293 | ||
| AK293172.1 | 15 | 2190 | ||
| AK314575.1 | ||||
| AY007130.1 | 7 | 1085 | AAG01994.1 | |
| BC009398.2 | 9 | 1216 | AAH09398.1 | |
| BC013375.2 | 15 | 2224 | AAH13375.1 | |
| D55716.1 | 15 | 2258 | BAA09534.1 | |
| D86748.1 | 15 | 2228 | ||
| KU178172.1 | ||||
| KU178173.1 | ||||
| X74796.1 | 12 | 1838 | CAA52803.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv607931 | Chr.7:100067194 - 100198523 on Build GRCh38 | Loss |
|
| nsv469687 | Chr.7:99974920 - 100157518 on Build GRCh38 | Loss |
|
| esv2734888 | Chr.7:99975623 - 101290285 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map