Assay Details
Target Gene Details
Entrez Gene ID: | 5914 |
Gene Name: | retinoic acid receptor alpha |
Gene Aliases: |
NR1B1, RAR |
Location: |
Chr.17:40309171-40357643 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RARA | NM_000964.3 | NP_000955.1 | ||
| NM_001145301.2 | 1 | 44 | NP_001138773.1 | |
| NM_001145302.2 | 1 | 44 | NP_001138774.1 | |
| XM_011525095.1 | XP_011523397.1 | |||
| AK130192.1 | ||||
| AK292205.1 | ||||
| AK312564.1 | ||||
| AL522713.3 | ||||
| BC008727.2 | AAH08727.1 | |||
| BG394590.1 | 1 | 45 | ||
| DA061829.1 | ||||
| DA541711.1 | ||||
| FJ487576.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833445 | Chr.17:40310222 - 40481717 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map