Assay Details
Target Gene Details
Entrez Gene ID: | 51294 |
Gene Name: | protocadherin 12 |
Gene Aliases: |
VE-cadherin-2, VECAD2 |
Location: |
Chr.5:141943585-141959062 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PCDH12 | NM_016580.3 | 4 | 4630 | NP_057664.1 |
| AB026893.1 | 4 | 3476 | BAA95162.1 | |
| AF231025.1 | 4 | 3725 | AAF61931.1 | |
| AF240635.1 | 4 | 4630 | AAF73962.1 | |
| AK023785.1 | 4 | 2293 | BAB14677.1 | |
| AK024140.1 | 4 | 3768 | BAB14837.1 | |
| AK027282.1 | BAB55016.1 | |||
| AK226049.1 | 4 | 3727 | ||
| AK315029.1 | 4 | 3727 | ||
| AY358428.1 | 4 | 3747 | AAQ88794.1 | |
| BC042634.1 | 4 | 3680 | AAH42634.1 | |
| BC052973.1 | 4 | 3737 | AAH52973.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1154058 | Chr.5:141632251 - 142199943 on Build GRCh38 | Deletion |
|
| nsv599924 | Chr.5:141919412 - 141955933 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map