Assay Details
Target Gene Details
Entrez Gene ID: | 338567 |
Gene Name: | potassium two pore domain channel subfamily K member 18 |
Gene Aliases: |
K2p18.1, MGR13, TRESK, TRESK-2, TRESK2, TRIK |
Location: |
Chr.10:117197489-117210299 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNK18 | NM_181840.1 | NP_862823.1 | ||
| AB087138.1 | BAC78527.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3624674 | Chr.10:117141857 - 117425204 on Build GRCh38 | Gain |
|
| nsv832002 | Chr.10:117037559 - 117246329 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map