Assay Details
Target Gene Details
Entrez Gene ID: | 2059 |
Gene Name: | epidermal growth factor receptor pathway substrate 8 |
Gene Aliases: |
DFNB102 |
Location: |
Chr.12:15620141-15789576 on Build GRCh38 |
Assay Gene Location: | Within Exon 22 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EPS8 | NM_004447.5 | 21 | 3476 | NP_004438.3 |
| XM_011520605.2 | 22 | 3536 | XP_011518907.1 | |
| BC030010.2 | 21 | 3288 | AAH30010.1 | |
| U12535.1 | 21 | 3248 | AAA62280.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs113738793] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map