Assay Details
Target Gene Details
Entrez Gene ID: | 344752 |
Gene Name: | arylacetamide deacetylase like 2 |
Gene Aliases: |
- |
Location: |
Chr.3:151733916-151758016 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AADACL2 | NM_207365.3 | 4 | 599 | NP_997248.2 |
| XM_011512753.1 | 4 | 627 | XP_011511055.1 | |
| BC065724.1 | 4 | 592 | AAH65724.1 | |
| BX647585.1 | 3 | 376 | CAI46075.1 | |
| CU449274.1 | 4 | 421 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv522472 | Chr.3:151743597 - 151864407 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map