Assay Details
Target Gene Details
Entrez Gene ID: | 162394 |
Gene Name: | schlafen family member 5 |
Gene Aliases: |
- |
Location: |
Chr.17:35243026-35273650 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLFN5 | NM_144975.3 | 2 | 1070 | NP_659412.3 |
| XM_005257934.3 | 2 | 1111 | XP_005257991.1 | |
| AK054668.1 | 2 | 1065 | BAB70788.1 | |
| AK303299.1 | 2 | 1065 | ||
| AK303632.1 | 2 | 1067 | ||
| BC021238.1 | 2 | 1048 | AAH21238.1 | |
| BC125200.1 | 1 | 1006 | ||
| BC125201.1 | 1 | 1006 | ||
| BX647942.1 | 2 | 1067 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2422399 | Chr.17:35212221 - 35291582 on Build GRCh38 | Deletion |
|
| esv3892998 | Chr.17:35106468 - 35295755 on Build GRCh38 | Loss |
|
| esv3892999 | Chr.17:35252863 - 35925533 on Build GRCh38 | Loss |
|
| nsv574741 | Chr.17:35225256 - 35500115 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map