Assay Details
Target Gene Details
Entrez Gene ID: | 56890 |
Gene Name: | Mdm1 nuclear protein |
Gene Aliases: |
- |
Location: |
Chr.12:68294566-68332381 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 14 - Exon 14 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MDM1 | NM_001205028.1 | NP_001191957.1 | ||
| NM_017440.4 | NP_059136.2 | |||
| XM_005269026.1 | XP_005269083.1 | |||
| XM_017019646.1 | XP_016875135.1 | |||
| XM_017019647.1 | XP_016875136.1 | |||
| XM_017019648.1 | XP_016875137.1 | |||
| XM_017019649.1 | XP_016875138.1 | |||
| XM_017019650.1 | XP_016875139.1 | |||
| AF007130.1 | AAC19149.1 | |||
| AK026745.1 | ||||
| AK297311.1 | ||||
| BC028355.2 | AAH28355.1 |
More Information
Set Membership: |
|

Genomic Map