Assay Details
Target Gene Details
Entrez Gene ID: | 51686 |
Gene Name: | ornithine decarboxylase antizyme 3 |
Gene Aliases: |
AZ3, OAZ-t, TISP15 |
Location: |
Chr.1:151762969-151771330 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OAZ3 | NM_001134939.1 | 3 | 384 | NP_001128411.1 |
| NM_001301371.1 | 3 | 271 | NP_001288300.1 | |
| NM_016178.2 | 3 | 419 | NP_057262.2 | |
| AF175296.1 | 3 | 419 | AAD51734.1 | |
| AK125876.1 | ||||
| AY713300.1 | 3 | 358 | ||
| BC073949.1 | 3 | 383 | AAH73949.1 | |
| BI830988.1 | 3 | 271 |
More Information
Set Membership: |
|

Genomic Map