Assay Details
Target Gene Details
Entrez Gene ID: | 81608 |
Gene Name: | factor interacting with PAPOLA and CPSF1 |
Gene Aliases: |
FIP1, Rhe, hFip1 |
Location: |
Chr.4:53377572-53462611 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
FIP1L1 | XM_005265769.4 | 19 | 2769 | XP_005265826.1 |
XM_005265773.4 | 18 | 2702 | XP_005265830.1 | |
XM_005265774.4 | 17 | 2614 | XP_005265831.1 | |
XM_005265778.4 | 18 | 2540 | XP_005265835.1 | |
XM_005265779.4 | 17 | 2468 | XP_005265836.1 | |
XM_005265781.4 | 17 | 2586 | XP_005265838.1 | |
XM_005265782.4 | 16 | 2505 | XP_005265839.1 | |
XM_017008662.1 | 17 | 2647 | XP_016864151.1 | |
XM_017008663.1 | 18 | 2603 | XP_016864152.1 | |
XM_017008664.1 | 17 | 2658 | XP_016864153.1 | |
XM_017008665.1 | 17 | 2574 | XP_016864154.1 | |
XM_017008666.1 | 16 | 2591 | XP_016864155.1 | |
XM_017008667.1 | 17 | 2557 | XP_016864156.1 | |
XM_017008668.1 | 16 | 2603 | XP_016864157.1 | |
XM_017008669.1 | 16 | 2503 | XP_016864158.1 | |
XM_017008670.1 | 16 | 2399 | XP_016864159.1 | |
XM_017008671.1 | 16 | 2318 | XP_016864160.1 | |
AK090938.1 | 7 | 1469 |
Target Gene Details
Entrez Gene ID: | 84708 |
Gene Name: | ligand of numb-protein X 1 |
Gene Aliases: |
LNX, MPDZ, PDZRN2 |
Location: |
Chr.4:53458264-53652524 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
LNX1 | XM_005265785.4 | 11 | 3384 | XP_005265842.1 |
XM_017008776.1 | 12 | 3445 | XP_016864265.1 | |
AF237782.1 | 10 | 2888 | AAK15039.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
