Assay Details
Target Gene Details
Entrez Gene ID: | 4524 |
Gene Name: | methylenetetrahydrofolate reductase (NAD(P)H) |
Gene Aliases: |
- |
Location: |
Chr.1:11785730-11806103 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
---|---|---|---|---|
MTHFR | NM_005957.4 | 12 | 2622 | NP_005948.3 |
XM_005263458.3 | 12 | 3459 | XP_005263515.1 | |
XM_005263460.4 | 12 | 3077 | XP_005263517.1 | |
XM_005263462.4 | 12 | 2510 | XP_005263519.1 | |
XM_005263463.3 | 12 | 2639 | XP_005263520.1 | |
XM_011541495.2 | 12 | 3456 | XP_011539797.1 | |
XM_011541496.2 | 12 | 3277 | XP_011539798.1 | |
AB209113.1 | BAD92350.1 | |||
BC053509.1 | 12 | 2444 | AAH53509.1 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
