Assay Details
Target Gene Details
Entrez Gene ID: | 7450 |
Gene Name: | von Willebrand factor |
Gene Aliases: |
F8VWF, VWD |
Location: |
Chr.12:5948874-6124675 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 38 - Exon 38 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| VWF | NM_000552.4 | NP_000543.2 | ||
| AK297600.1 | ||||
| AK298097.1 | ||||
| AK301216.1 | ||||
| M10321.1 | AAB59512.1 | |||
| X04385.1 | CAA27972.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv557191 | Chr.12:5972658 - 6106468 on Build GRCh38 | Loss |
|
| nsv468988 | Chr.12:5413390 - 6141521 on Build GRCh38 | Gain |
|
| dgv447e212 | Chr.12:5987747 - 5992371 on Build GRCh38 | Loss |
|
| esv3580088 | Chr.12:5598006 - 6123173 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map