Assay Details
Target Gene Details
Entrez Gene ID: | 79661 |
Gene Name: | nei like DNA glycosylase 1 |
Gene Aliases: |
FPG1, NEI1, hFPG1 |
Location: |
Chr.15:75346895-75355251 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2621n100 | Chr.15:75101872 - 75382048 on Build GRCh38 | Gain |
|
| esv2749894 | Chr.15:75326873 - 75749895 on Build GRCh38 | Deletion |
|
| nsv1615 | Chr.15:75351396 - 75377578 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map